Learn More
Medchemexpress LLC Yohimban-16-carboxylic acid, 11,17-dimethoxy-18-[[3,4,5-tri(methoxy-d3)benzoyl]oxy]-, methyl ester, (3β,163,17α,18β,20α)- | 84759-11-5 | 617.73 | 1 MG
SDP
Catalog No. 5000389077
Encompass_Preferred
Reserpine-d9 is the deuterium labeled Reserpine. Reserpine is an inhibitor of the vesicular monoamine transporter 2 (VMAT2). Deuteration can affect the pharmacokinetic and metabolic profiles of drugs. This compound is for research use only, not sold to patients.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-89077 Supplier Medchemexpress LLC Supplier No. HYN0480S1MG
Reserpine-d9 is the deuterium labeled Reserpine. Reserpine is an inhibitor of the vesicular monoamine transporter 2 (VMAT2). Deuteration can affect the pharmacokinetic and metabolic profiles of drugs. This compound is for research use only, not sold to patients.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.