Learn More
Medchemexpress LLC Yohimbine-d3 | 133146-00-6 | 99.0% | 357.46 g/mol | C21H23D3N2O3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY12715S15MG
Encompass_Preferred
Yohimbine-d3 is the deuterium-labeled analogue of yohimbine intended for use as a tracer or internal standard in analytical and mass-spectrometry workflows. It provides a mass-differentiated reference to support accurate quantitation and method validation.
Catalog No. 50-002-64106