Learn More
Medchemexpress LLC Yohimbine-d3 | 133146-00-6 | 99.36% | 1 MG
SDP
Catalog No. 5000342417
Encompass_Preferred
Yohimbine-d3 is a deuterium-labeled version of Yohimbine (HY-12715). This compound serves as a tracer and can be used as an internal standard for quantitative analysis through techniques like NMR, GC-MS, or LC-MS. Deuterium substitution in drug molecules is being explored for its potential to influence pharmacokinetic and metabolic profiles.
  • Deuterium labeled yohimbine.
  • Can be used as a tracer.
  • Suitable as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration may impact pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-42417 Supplier Medchemexpress LLC Supplier No. HY12715S11MG
Yohimbine-d3 is a deuterium-labeled version of Yohimbine (HY-12715). This compound serves as a tracer and can be used as an internal standard for quantitative analysis through techniques like NMR, GC-MS, or LC-MS. Deuterium substitution in drug molecules is being explored for its potential to influence pharmacokinetic and metabolic profiles.
  • Deuterium labeled yohimbine.
  • Can be used as a tracer.
  • Suitable as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration may impact pharmacokinetic and metabolic profiles of drugs.