Learn More
AdipoGen Yohimbine hydrochloride
SDP
Supplier:  AdipoGen CDXY0013G005
Encompass_Preferred
Chemical. CAS 65-19-0. Formula C21H26N2O3 . HCl. MW 354.4 . 36.5. Synthetic. alpha2-Adrenoceptor antagonist. pKi values are 8.52, 8.00 and 9.17 for human alpha2A, alpha2B and alpha2C receptors, respectively.
Catalog No. 50-255-9897