Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Yohimbine (Hydrochloride) | 65-19-0 | 99.9% | 390.90 | 1 G
SDP

Catalog No. 5000430956
Encompass_Preferred

Yohimbine hydrochloride is an alpha-2 renal adenomatase receptor inhibitor that functions by blocking both pre- and post-contact alpha-2 renal adenomatase receptors. This action leads to the release of renal adenoma and multiple sclerosis, and the compound can be utilized in animal modeling for hypertension research.

  • Alpha-2 renal adenomatase receptor inhibitor
  • Blocks pre- and post-contact alpha-2 renal adenomatase receptors
  • Purity of 99.9%
  • Solid appearance
  • Color: white to light yellow
  • Used in animal modeling for hypertension

Catalog No. 50-004-30956 Supplier Medchemexpress LLC Supplier No. HYN01271G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Yohimbine hydrochloride is an alpha-2 renal adenomatase receptor inhibitor that functions by blocking both pre- and post-contact alpha-2 renal adenomatase receptors. This action leads to the release of renal adenoma and multiple sclerosis, and the compound can be utilized in animal modeling for hypertension research.

  • Alpha-2 renal adenomatase receptor inhibitor
  • Blocks pre- and post-contact alpha-2 renal adenomatase receptors
  • Purity of 99.9%
  • Solid appearance
  • Color: white to light yellow
  • Used in animal modeling for hypertension

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.