Learn More
Cayman Chemical Yohimbine (hydrochloride), Purity: ≥98%, CAS: 65-19-0, 10 g
SDP
Catalog No. 502984933
Encompass_Preferred
A natural alkaloid that has antagonist activity at α2-adrenergic receptors (pKis = 8.2, 8.7, and 9.6 for human α2A, α2B, and α2C, respectively
Catalog No. 50-298-4933 Supplier Cayman Chemical Supplier No. 1986910 G
A natural alkaloid that has antagonist activity at α2-adrenergic receptors (pKis = 8.2, 8.7, and 9.6 for human α2A, α2B, and α2C, respectively