Learn More
Medchemexpress LLC YOK-2204 | 2409959-99-3 | 98.6% | 449.58 | 25 MG
SDP
Catalog No. 5000391393
Encompass_Preferred
YOK-2204 is a ligand of the p62-ZZ domain. It can activate p62-dependent selective macroautophagy and can be utilized for AUTOTAC design.
  • Ligand of p62-ZZ domain.
  • Activates p62-dependent selective macroautophagy.
  • Can be used for AUTOTAC design.
Catalog No. 50-003-91393 Supplier Medchemexpress LLC Supplier No. 25MGHY161738
YOK-2204 is a ligand of the p62-ZZ domain. It can activate p62-dependent selective macroautophagy and can be utilized for AUTOTAC design.
  • Ligand of p62-ZZ domain.
  • Activates p62-dependent selective macroautophagy.
  • Can be used for AUTOTAC design.