Learn More
Grainger Smith Bearing Flanged Yoke Roller, Alloy Steel, 5 in. Diameter, 1 ea
Catalog No. 502760242
Encompass_Preferred
This heavy-duty flanged yoke roller features a robust alloy steel construction, designed to accommodate significant radial and thrust load applications. Utilizing a double-row tapered roller bearing element, this unit is built for reliability in demanding industrial environments.
  • Bore diameter: 1.75 in. (4.45 cm)
  • Roller diameter: 5 in. (12.7 cm)
  • Temperature range: -40°F to 250°F
  • Double sealed enclosure for contamination resistance
  • Large lubrication reservoir for extended service life
  • Black oxide finish coating
  • Relubricatable design
Catalog No. 50-276-0242 Supplier Grainger Supplier No. 15U601
This heavy-duty flanged yoke roller features a robust alloy steel construction, designed to accommodate significant radial and thrust load applications. Utilizing a double-row tapered roller bearing element, this unit is built for reliability in demanding industrial environments.
  • Bore diameter: 1.75 in. (4.45 cm)
  • Roller diameter: 5 in. (12.7 cm)
  • Temperature range: -40°F to 250°F
  • Double sealed enclosure for contamination resistance
  • Large lubrication reservoir for extended service life
  • Black oxide finish coating
  • Relubricatable design