Learn More
Apexbio Technology LLC YONKENAFIL-HYDROCHLORIDE-1MG
SDP
Catalog No. 5000567868
Encompass_Preferred
Yonkenafil hydrochloride (CAS No 804519-64-0) is a compound extensively studied within the field of biomedical research for its potential therapeutic effects Originating as a research chemical it is known to act on phosphodiesterase (PDE) pathways particularly targeting the PDE5 enzyme which plays a significant role in vasodilation processes Its in vitro activity is often noted in the nanomolar to low micromolar range making it suitable for potent pharmacological interventions Typical applications include use in various cell models and animal studies to explore cardiovascular or reproductive health implications as well as in drug screening protocols to assess efficacy and safety Experimental concentrations typically depend on the specific research design allowing for flexible integration into different experimental frameworks
Catalog No. 50-005-67868 Supplier Apexbio Technology LLC Supplier No. C86921
Yonkenafil hydrochloride (CAS No 804519-64-0) is a compound extensively studied within the field of biomedical research for its potential therapeutic effects Originating as a research chemical it is known to act on phosphodiesterase (PDE) pathways particularly targeting the PDE5 enzyme which plays a significant role in vasodilation processes Its in vitro activity is often noted in the nanomolar to low micromolar range making it suitable for potent pharmacological interventions Typical applications include use in various cell models and animal studies to explore cardiovascular or reproductive health implications as well as in drug screening protocols to assess efficacy and safety Experimental concentrations typically depend on the specific research design allowing for flexible integration into different experimental frameworks