Learn More
Medchemexpress LLC Yonkenafil hydrochloride | 804519-64-0 | 99.4% | 524.08 | C24H34ClN5O4S | 100MG
SDP
Catalog No. 5000238117
Encompass_Preferred
Yonkenafil hydrochloride is a phosphodiesterase 5 (PDE5) inhibitor provided as the hydrochloride salt for preclinical research. It has been reported to reduce cerebral infarction and neuronal damage and to modulate neurogenesis, with formulation guidance for both in vitro and in vivo use.
Catalog No. 50-002-38117 Supplier Medchemexpress LLC Supplier No. HY125095100MG
Yonkenafil hydrochloride is a phosphodiesterase 5 (PDE5) inhibitor provided as the hydrochloride salt for preclinical research. It has been reported to reduce cerebral infarction and neuronal damage and to modulate neurogenesis, with formulation guidance for both in vitro and in vivo use.