Learn More
Medchemexpress LLC Yonkenafil hydrochloride | 804519-64-0 | 99.4% | 524.08 | C24H34ClN5O4S | 10MG
SDP
Catalog No. 5000238689
Encompass_Preferred
Yonkenafil hydrochloride is a research-grade phosphodiesterase 5 (PDE5) inhibitor supplied as the hydrochloride salt for biochemical and pharmacological studies. It is commonly used in in vitro and in vivo experiments to investigate PDE5-mediated signaling, vascular effects, and neuroprotection, and is typically handled as a DMSO-soluble compound.
  • Research-grade PDE5 inhibitor for biochemical and pharmacological studies.
  • High purity suitable for sensitive assays.
  • Hydrochloride salt form improves solubility in polar solvents.
  • Soluble in DMSO for easy assay preparation.
  • Reported molecular formula C24H34ClN5O4S and molecular weight 524.08.
  • Available in multiple small pack sizes for laboratory use.
Catalog No. 50-002-38689 Supplier Medchemexpress LLC Supplier No. HY12509510MG
Yonkenafil hydrochloride is a research-grade phosphodiesterase 5 (PDE5) inhibitor supplied as the hydrochloride salt for biochemical and pharmacological studies. It is commonly used in in vitro and in vivo experiments to investigate PDE5-mediated signaling, vascular effects, and neuroprotection, and is typically handled as a DMSO-soluble compound.
  • Research-grade PDE5 inhibitor for biochemical and pharmacological studies.
  • High purity suitable for sensitive assays.
  • Hydrochloride salt form improves solubility in polar solvents.
  • Soluble in DMSO for easy assay preparation.
  • Reported molecular formula C24H34ClN5O4S and molecular weight 524.08.
  • Available in multiple small pack sizes for laboratory use.