Learn More
Medchemexpress LLC Yonkenafil hydrochloride | 804519-64-0 | 99.4% | 524.08 g/mol | C24H34ClN5O4S | 25MG
SDP
Catalog No. 5000237543
Encompass_Preferred
Yonkenafil hydrochloride is a research-grade phosphodiesterase 5 (PDE5) inhibitor intended for preclinical and biochemical studies. The compound is supplied as a hydrochloride salt, with molecular formula C24H34ClN5O4S and molecular weight 524.08 g/mol. It is typically used as a reference or test compound in pharmacology and neurobiology research where PDE5 modulation is of interest.
  • PDE5 inhibitor suitable for biochemical and cellular assays.
  • Reported high purity (99.4%) for consistent experimental results.
  • Available in small milligram pack sizes for assay development.
  • Hydrochloride salt form for improved handling and stability.
  • Molecular properties provided for formulation and dosing calculations.
Catalog No. 50-002-37543 Supplier Medchemexpress LLC Supplier No. HY12509525MG
Yonkenafil hydrochloride is a research-grade phosphodiesterase 5 (PDE5) inhibitor intended for preclinical and biochemical studies. The compound is supplied as a hydrochloride salt, with molecular formula C24H34ClN5O4S and molecular weight 524.08 g/mol. It is typically used as a reference or test compound in pharmacology and neurobiology research where PDE5 modulation is of interest.
  • PDE5 inhibitor suitable for biochemical and cellular assays.
  • Reported high purity (99.4%) for consistent experimental results.
  • Available in small milligram pack sizes for assay development.
  • Hydrochloride salt form for improved handling and stability.
  • Molecular properties provided for formulation and dosing calculations.