Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Yonkenafil hydrochloride | 804519-64-0 | 99.4% | 524.08 g/mol | C24H34ClN5O4S | 50MG
SDP

Catalog No. 5000238589
Encompass_Preferred

Yonkenafil hydrochloride is a research-grade phosphodiesterase 5 (PDE5) inhibitor supplied as the hydrochloride salt for laboratory pharmacology studies. It has demonstrated neuroprotective and cognitive effects in preclinical models and is suitable for both in vitro assays and in vivo formulations. This material is intended for research use only and should be handled and stored to minimize moisture exposure.

  • Purity 99.4%.
  • Molecular weight 524.08 g/mol.
  • Chemical formula C24H34ClN5O4S.
  • Soluble in DMSO at 100 mg/mL with sonication.
  • Formulatable for in vivo dosing at ≥2.5 mg/mL in recommended vehicles.
  • Store sealed and protected from moisture and light; freeze solutions for longer-term storage.

Catalog No. 50-002-38589 Supplier Medchemexpress LLC Supplier No. HY12509550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Yonkenafil hydrochloride is a research-grade phosphodiesterase 5 (PDE5) inhibitor supplied as the hydrochloride salt for laboratory pharmacology studies. It has demonstrated neuroprotective and cognitive effects in preclinical models and is suitable for both in vitro assays and in vivo formulations. This material is intended for research use only and should be handled and stored to minimize moisture exposure.

  • Purity 99.4%.
  • Molecular weight 524.08 g/mol.
  • Chemical formula C24H34ClN5O4S.
  • Soluble in DMSO at 100 mg/mL with sonication.
  • Formulatable for in vivo dosing at ≥2.5 mg/mL in recommended vehicles.
  • Store sealed and protected from moisture and light; freeze solutions for longer-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.