Learn More
Eurofins Discoverx YopH, active, 250 µg. (14-590M)
Supplier:  Eurofins Discoverx 14590M
Encompass
The YopH, active product is purified N terminal GST-tagged recombinant YopH enzyme. This enzyme is full length. It retains its binding and enzymatic activity and is intended for use in phosphatase assays. This enzyme is provided as 250 μg of 3.31 mg/mL frozen solution. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9014