Learn More
AURORA BIOLABS LLC YopH (CLP), 1000 units
Catalog No. NC3787973
Encompass
The PTP catalytic domain (a.a. 163 to a.a. 468) of Yersinia enterocolitica YopH is expressed in E.coli. The tyrosine-protein phosphatase YopH selectively hydrolyzes only phosphortyrosine-containing proteins and contains a Cys residue essential for catalysis.
Catalog No. NC3787973 Supplier AURORA BIOLABS LLC Supplier No. 200100100
The PTP catalytic domain (a.a. 163 to a.a. 468) of Yersinia enterocolitica YopH is expressed in E.coli. The tyrosine-protein phosphatase YopH selectively hydrolyzes only phosphortyrosine-containing proteins and contains a Cys residue essential for catalysis.