Learn More
MSC Youngstown Arc Flash Gloves, 20.8 cal/cm2, Goatskin, Medium, 1 Pair
Catalog No. 5000687531
Encompass
These arc flash gloves are designed for superior protection and durability in hazardous environments. Made from goatskin leather, they offer a high level of arc flash protection.
  • Double layer leather
  • Incredible durability
  • Unlined
  • Maximum arc flash protection rating: 20.8 cal/cm²
  • NFPA 70E compliant
Catalog No. 50-006-87531 Supplier MSC Supplier No. 51639946
These arc flash gloves are designed for superior protection and durability in hazardous environments. Made from goatskin leather, they offer a high level of arc flash protection.
  • Double layer leather
  • Incredible durability
  • Unlined
  • Maximum arc flash protection rating: 20.8 cal/cm²
  • NFPA 70E compliant