Learn More
MSC Youngstown Arc Flash Gloves, 20.8 cal/cm2, Goatskin, Small, 1 Pair
Catalog No. 5000794818
Encompass
The Leather Utility Plus gloves are climbing gloves for electric utility workers. They feature specially tanned goatskin grain leather for grip and abrasion resistance, an ergonomic 3D pattern for comfort and dexterity, and are arc rated for protection.
  • Goatskin grain leather for strong grip and abrasion resistance
  • Ergonomic 3D pattern for comfort and dexterity
  • High durability; outlasts traditional leather gloves
  • Double layer leather on fingertips, knuckle, saddle, and palm
  • Features a 4-inch safety cuff for extended wrist protection
  • Resistant to dripping, melting, or ignition from sparks or flames
  • Arc rating: 20.8 cal/cm² (ASTM 2675)
  • Puncture resistance: ANSI/ISEA 105 Level 4
  • Complies with NFPA 70e-2018 and OSHA 29 CFR Part 1926
Catalog No. 50-007-94818 Supplier MSC Supplier No. 51639953
The Leather Utility Plus gloves are climbing gloves for electric utility workers. They feature specially tanned goatskin grain leather for grip and abrasion resistance, an ergonomic 3D pattern for comfort and dexterity, and are arc rated for protection.
  • Goatskin grain leather for strong grip and abrasion resistance
  • Ergonomic 3D pattern for comfort and dexterity
  • High durability; outlasts traditional leather gloves
  • Double layer leather on fingertips, knuckle, saddle, and palm
  • Features a 4-inch safety cuff for extended wrist protection
  • Resistant to dripping, melting, or ignition from sparks or flames
  • Arc rating: 20.8 cal/cm² (ASTM 2675)
  • Puncture resistance: ANSI/ISEA 105 Level 4
  • Complies with NFPA 70e-2018 and OSHA 29 CFR Part 1926