Learn More
MSC Youngstown Arc Flash Gloves, 3X-Large, 23 cal/cm2, Goatskin, 1 Pair
Catalog No. 5000686175
Encompass
These arc flash gloves are constructed from goatskin leather and provide robust protection for arc flash hazards, making them suitable for demanding industrial applications.
  • Maximum arc flash protection rating of 23 cal/cm²
  • Size 3X-Large for optimal fit
  • Constructed with goatskin leather
  • Double-layer leather for incredible durability
  • Unlined for enhanced dexterity
  • Meets ASTM F2675 standards
  • ANSI/ISEA abrasion resistance level 2
  • ANSI/ISEA cut resistance level 2
Catalog No. 50-006-86175 Supplier MSC Supplier No. 66802695
These arc flash gloves are constructed from goatskin leather and provide robust protection for arc flash hazards, making them suitable for demanding industrial applications.
  • Maximum arc flash protection rating of 23 cal/cm²
  • Size 3X-Large for optimal fit
  • Constructed with goatskin leather
  • Double-layer leather for incredible durability
  • Unlined for enhanced dexterity
  • Meets ASTM F2675 standards
  • ANSI/ISEA abrasion resistance level 2
  • ANSI/ISEA cut resistance level 2