Learn More
MSC Youngstown Arc Flash Gloves, Medium, 23 cal/cm2, 1 Pair
Catalog No. 5000673928
Encompass
These arc flash gloves are designed for protection against arc flash hazards. They are unlined and conform to the ASTM F2675 standard, offering reliable safety in demanding environments.
  • Maximum arc flash protection rating of 23 cal/cm2
  • Unlined for enhanced dexterity
  • Conforms to ASTM F2675 standard
  • ANSI/ISEA Abrasion Resistance Level 2
  • ANSI/ISEA Cut Resistance Level 2
  • Size medium for comfortable fit
  • Supplied as 1 pair
Catalog No. 50-006-73928 Supplier MSC Supplier No. 66802752
These arc flash gloves are designed for protection against arc flash hazards. They are unlined and conform to the ASTM F2675 standard, offering reliable safety in demanding environments.
  • Maximum arc flash protection rating of 23 cal/cm2
  • Unlined for enhanced dexterity
  • Conforms to ASTM F2675 standard
  • ANSI/ISEA Abrasion Resistance Level 2
  • ANSI/ISEA Cut Resistance Level 2
  • Size medium for comfortable fit
  • Supplied as 1 pair