Learn More
MSC Youngstown Arc Flash Gloves, X-Large Goatskin, 12 cal, 1 ea
Catalog No. 5000794817
Encompass
These Arc Flash Gloves provide essential protection for industrial applications, featuring an ergonomic design for comfort and exceptional durability. With a 12 cal/cm² rating, they are constructed from goatskin leather and lined with KEVLAR® for enhanced safety.
  • Ergonomic design
  • Incredibly durable
  • Goatskin leather construction
  • Kevlar® lining
  • Arc flash protection, 12 cal/cm²
  • Meets ASTM F2302 and NFPA 70E standards
  • ANSI/ISEA abrasion resistance level 3
  • ANSI/ISEA cut resistance level 3 (A4)
Catalog No. 50-007-94817 Supplier MSC Supplier No. 51639813
These Arc Flash Gloves provide essential protection for industrial applications, featuring an ergonomic design for comfort and exceptional durability. With a 12 cal/cm² rating, they are constructed from goatskin leather and lined with KEVLAR® for enhanced safety.
  • Ergonomic design
  • Incredibly durable
  • Goatskin leather construction
  • Kevlar® lining
  • Arc flash protection, 12 cal/cm²
  • Meets ASTM F2302 and NFPA 70E standards
  • ANSI/ISEA abrasion resistance level 3
  • ANSI/ISEA cut resistance level 3 (A4)