Learn More
Youngstown Barrel & Drum Ultra-Absorbent Tarp™

Description
Catch and trap oil drips before they can reach the ground.
- Rolls or folds into small package — stores in vehicles for onsite repairs
- Keep floors free of oily mess — reduce slip hazards
- Top layer: UV resistant geotextile fabric that allows oils to pass through
- Middle layer: Absorbent core; oil-only polypropylene traps oils but allows stormwater to evaporate
- Bottom layer: Impermeable backing keeps drips from leaching through to the ground or shop floor
Compliance
Helps comply with NPDES, 40 CFR 122.26 (1999)
Specifications
Specifications
| Certifications/Compliance | NPDES, 40 CFR 122.26 |
| Capacity (English) | 3 gal. |
| Capacity (Metric) | 11.3 L |
| Type | Absorbent Tarp |
| For Use With (Application) | Use under machinery and equipment to catch fluid leaks during maintenance or repairs, keeps floors free of oily mess reduces slip hazards |
| For Use With (Equipment) | Any machinery and equipment |
| Includes | UV-resistant geotextile fabric, oil, impermeable backing |
| Dimensions (L x W) | 5 x 5 ft. (152.4 x 152.4 cm) |
| Length (English) | 5 ft. |
| Length (Metric) | 152.4 cm |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.