Learn More
Youngstown Barrel & Drum Ultra-Scissor Lift Diaper™

Description
Includes
Eight fastening magnets
- Keeps hydraulic oil and other lubricants from dripping onto floors, eliminating slip hazards and messy clean-ups
- Absorbent material is poly-backed to keep absorbed liquids from leaking through
- Stays in place even when equipment is moved to different locations
- Dimensions: 3L × 5mW (10 × 16 ft.)
Specifications
Specifications
| Type | Ultra Scissor Lift Diaper |
| For Use With (Application) | Absorbs oils and lubricant |
| Includes | Fastening magnets |
| Dimensions (L x W) | 10 ft. x 16 in. (3m x 40.6 cm) |
| Length (English) | 10 ft. |
| Length (Metric) | 3 m |
| Width (English) | 16 in. |
| Width (Metric) | 40.6 cm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.