Learn More
Youngstown Barrel & Drum Ultra-Drum Tourniquets™
SDP
Catalog No. 19339027
Change view
Click to view available options
Weight (English):
5 lb.
9 lb.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Weight (English)
19-339-027 5 lb.
19-820-927 9 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-339-027 Supplier Youngstown Barrel & Drum Supplier No. 2010 UT
Includes
UV-resistant bag included
Latches onto drums to seal off leaks
A chemical-resistant sealing pad is compressed against the damaged area to stop leaks on contact.
  • 8.9x17.8cm (3.5x7 in.) maximum sealing area
  • 305cm (10 ft.) long ratchet strap
  • No tools required
  • Magnetic model has powerful magnets that latch onto steel drums to seal off puncture and corrosion leaks. A chemical-resistant sealing pad is compressed against the damaged area to stop leaks on contact
  • Universal model features a ratchet strap system. Works on all surfaces including polyethylene, steel and stainless steel
For temporary use only
Specifications
Product Type Magnetic Drum Tourniquet
For Use With (Application) Seal off puncture and corrosion leaks on drums and works on all surfaces including Polyethylene, Steel and Stainless Steel
For Use With (Equipment) 8.9 x 17.8 cm Steel Drums
Includes Stainless Steel plate, chemical resistant EDPM pad, UV-resistant bag, 2 magnets, spark resistant assembly
Weight (English) 5 lb.