Learn More
Youngstown Barrel & Drum Ultra-Sampling Kit™
SDP
Catalog No. 18938454
Change view
Click to view available options
Sampling Kit
Catalog No.
18-938-454 Sampling Kit
1 options
Catalog No. 18-938-454 Supplier Youngstown Barrel & Drum Supplier No. 9297 UT
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Includes
  • Turbidity meter
  • pH meter
  • pH and turbidity meter calibration solution
  • Durable case
  • Data book
  • Lint free wipes
  • Nitrile gloves
Stormwater sampling for pH and turbidity
Specifications
Product Type Sampling Kit
Includes Turbidity meter, pH meter, pH and turbidity meter calibration solution, durable case, data book, lint free wipes, nitrile gloves
Weight (English) 5 lb.
Weight (Metric) 2.3 Kg