Learn More
Youngstown Barrel & Drum Ultra-Bung Access Funnels™: Accessories
SDP
Catalog No. 191304823
Change view
Click to view available options
Pop-up fill gauge
Catalog No.
19-130-4823 Pop-up fill gauge
1 options
Catalog No. 19-130-4823 Supplier Youngstown Barrel & Drum Supplier No. 9985 UT
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
For use with Youngstown Barrel & Drum Ultra-Bung Access Funnels
For Ultra-Bung Access Funnels, see cat. no. 191300640
Specifications
Capacity (Metric) 22.7 L
For Use With (Application) Allows visual monitoring of waste level in the drum and helps plan ahead for drum removal
Diameter (Metric) Opening 60 cm