Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Youngstown Barrel & Drum Ultra-Drain Guard™ Curb Insert
SDP

Catalog No. 19339047
Change view
Click to view available options
For Use With (Equipment):
Grates 40 x 40 in., Curb Inlets to 60 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-339-047 Grates 40 x 40 in., Curb Inlets to 60 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-339-047 Supplier Youngstown Barrel & Drum Supplier No. 9239 UT

Filters contaminants from street grate/curb opening catch basins

Designed for use with catch basins that have a combination street grate and curb opening

  • Heavy-duty tension rod secures insert inside the curb inlet, blocking the passage of contaminants
  • Fits grate sizes up to 40 × 40 in. (102 × 102cm) and curb inlets up to 60 in. (152cm)
  • Oil and Sediment Plus insert features X-Tex™ filter strips to increase oil and grease absorption

Compliance

Considered a Best Management Practice in compliance with NPDES, 40CFR122.26 (1999).

Specifications

Certifications/Compliance 40 CFR 122.26
Type Drain Guard
For Use With (Application) Filters contaminants from street grate or curb opening catch basins
For Use With (Equipment) Grates 40 x 40 in., Curb Inlets to 60 in.
Includes X-Tex filter strips
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.