Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Youngstown Barrel & Drum DrumSaver Insert
SDP

Catalog No. 18942A
Change view
Click to view available options
Capacity (English):
55 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
18-942A 55 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-942A Supplier Youngstown Barrel & Drum Supplier No. 3140
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For 55 gal. (208L) open-head steel drums

  • 15mil LDPE
  • Corrugated, accordion style

Specifications

Product Type Drum Liner
Capacity (English) 55 gal.
Capacity (Metric) 78 L
For Use With (Application) Drumsaver Insert for Steel Drums
Material Fiber
Thickness (English) 15 mil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.