Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Youngstown Barrel & Drum Key for Lever Lock Ring
SDP

Catalog No. 19339744
Change view
Click to view available options
For Use With (Equipment):
Fiber Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-339-744 Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-339-744 Supplier Youngstown Barrel & Drum Supplier No. 2888

missing translation for 'validMainframeMsg'

Accessory for lever lock rings for drums.

Drum Key for lever lock rings.

Specifications

Product Type Lever Lock Key
Color Silver
For Use With (Application) Opening and closing a lever lock ring for a fiber drum
For Use With (Equipment) Fiber Drum
Material Steel
Weight (English) 0.5 lb.
Weight (Metric) 0.22679618 Kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.