Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Youngstown Barrel & Drum LDPE Drum Liners
SDP

Catalog No. 191314720
Change view
Click to view available options
Capacity (English):
55 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
19-131-4720 55 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-131-4720 Supplier Youngstown Barrel & Drum Supplier No. 3150
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Let's you reuse your drum.

LDPE Drum Liners. Drumsaver insert, Straight Sided, for open-head 55 gallon steel drums. 24 mil. Minimum 1.

Specifications

Product Type Drum Liner
Capacity (English) 55 gal.
Capacity (Metric) 78 L
Color Clear
For Use With (Application) Drumsaver insert, straight Sided, for open-head Steel drums
For Use With (Equipment) For 55 gal. Drums
Material LDPE
Thickness (English) 24 mil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.