Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Youngstown Barrel & Drum Pailsaver Dust Caps
SDP

Catalog No. 191314753
Change view
Click to view available options
For Use With (Equipment):
55 gal. drums
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-131-4753 55 gal. drums
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-131-4753 Supplier Youngstown Barrel & Drum Supplier No. 3189
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Elasticized cap protects open drums from contaminents.

LDPE Pailsaver Dust Caps for 55 gallon drums. 4 mil. MOQ of 100/ea.

Specifications

Color Clear
Capacity (English) 55 gal.
Capacity (Metric) 208.2 L
Material LDPE
Type Dust Cap
For Use With (Application) Cover 55 gal drums
For Use With (Equipment) 55 gal. drums
Packaging Quantity 100/Pk.
Diameter (English) 34 in.
Diameter (Metric) 86.36 cm
Weight (English) 0.12 lb.
Weight (Metric) 0.0544 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.