Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Youngstown Barrel & Drum 3.5 Gallon Plastic Pails, Open Head-New Generation
SDP

Catalog No. 19823260
Change view
Click to view available options
Capacity (English):
3.5 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
19-823-260 3.5 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-823-260 Supplier Youngstown Barrel & Drum Supplier No. 2379NG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Store or ship smaller quantities of solids.

3.5 Gallon Plastic Pails, Open Head-New Generation. Screw lid with Neoprene gasket. UN rated 1H2/Y19/S. White.

Specifications

Certifications/Compliance UN 1H2/Y19/S
Color White
Design Open head, removable lid
Capacity (English) 3.5 gal.
Capacity (Metric) 15.4 L
Material Plastic
Type Pail
For Use With (Application) Packaging for hazardous solids
Includes Screw lid with neoprene gasket
Model Life Latch
Packaging Quantity 1 Ea.
Shape Circular
Dimensions (L x W x H) 12.88 x 12.88 x 12.20 in.
Length (English) 12.88 in.
Length (Metric) 32.7152 cm
Width (English) 12.88 in.
Width (Metric) 32.7152 cm
Height (English) 12.88 in.
Height (Metric) 32.7152 cm
Depth (English) 11.125 in.
Depth (Metric) 28.2575 cm
Diameter (English) 11.25 in. I.D., 12.88 in. O.D.
Diameter (Metric) 28.575 cm I.D., 32 cm O.D.
Weight (English) 6.6 lb.
Weight (Metric) 3 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.