Learn More
Youngstown Barrel & Drum Ultra-Burp-Free Funnel™
Description
Rust-free, noncorroding polyethylene construction provides superior chemical resistance.
- Hinged lid may be locked to prevent unauthorized access
- With threaded brass insert screws to fit a 2 in. (5.1cm) drum bung
Specifications
Specifications
| Material | Polyethylene |
| Includes | Built-in vent, lockable hinged lid, threaded brass insert |
| Height (Metric) | 29.2 cm |