Learn More
Youngstown Barrel & Drum Ultra-Drum Tourniquets™
Description
Includes
UV-resistant bag included
A chemical-resistant sealing pad is compressed against the damaged area to stop leaks on contact.
- 8.9x17.8cm (3.5x7 in.) maximum sealing area
- 305cm (10 ft.) long ratchet strap
- No tools required
- Magnetic model has powerful magnets that latch onto steel drums to seal off puncture and corrosion leaks. A chemical-resistant sealing pad is compressed against the damaged area to stop leaks on contact
- Universal model features a ratchet strap system. Works on all surfaces including polyethylene, steel and stainless steel
For temporary use only
Specifications
Specifications
| Product Type | Universal Drum Tourniquet |
| For Use With (Application) | Seal off puncture and corrosion leaks on drums and works on all surfaces including Polyethylene, Steel and Stainless Steel |
| For Use With (Equipment) | Steel Drums |
| Includes | 10 ft. L Ratchet strap, Stainless Steel backing plate, chemical resistant EDPM sealing pad, UV-resistant bag, spark resistant assembly |
| Weight (English) | 9 lb. |