Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Youngstown Barrel & Drum Ultra-Overpack Plus™ 95 Poly Drum
SDP

Catalog No. 19171522
Change view
Click to view available options
Capacity (English):
65 gal.
95 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
19-171-522 95 gal.
19-824-100 65 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-171-522 Supplier Youngstown Barrel & Drum Supplier No. 0580 UT

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Handles damaged or leaking containers of hazardous materials

  • Open head style
  • Easy-to-close screw top lid
  • Color: Yellow
  • UN marking: 1H2\X345\S\09\USA\M5904

Order Info

Capacity: 95 gal. (360L). O.D. x H: 32.3 x 43.3 in. (82 x 110cm). Weight: 48 lb. (22kg).

Certifications

UN Transport of Dangerous Goods. IMO Dangerous Goods Code (IMDG) Annex I-Section 8. ICAO Technical Instructions for the Safe Transport of Dangerous Goods by Air, Part 7, Chapter 4. IATA Dangerous Goods Regulations, Section 10.3.

Compliance

Complies with UN packaging group 1 (X-Rated) and certified for use as DOT Salvage Drums, 49 CFR 173.3.

Specifications

Certifications/Compliance UN 1H2\X345\S\09\USA\M5904
Closure Type Screw Lid
Capacity (English) 95 gal.
Capacity (Metric) 360 L
Color Yellow
For Use With (Application) For handles damaged or leaking containers of hazardous materials
Material Plastic
Freight Class 50
Head Style Open
Height (English) 41.25 in.
Diameter (English) Outer 25.5 to 32.25 in.
Diameter (Metric) Outer 64.77 to 81.91 mm
Weight (English) 48 lb.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.