Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Youngstown Barrel & Drum Ultra-Decon Deck™, Hospital Model
SDP

Catalog No. 19339008
Change view
Click to view available options
Capacity (English):
176 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
19-339-008 176 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-339-008 Supplier Youngstown Barrel & Drum Supplier No. 6010 UT

Includes

  • DeconDeck, two bladder attachments, ground tarp: 12 x 14 ft. (3.66 x 4.27m), gross rinse shower unit, spray wand (7 gpm output), privacy shelter, supply hose

Easy to set up

 

  • Privacy shelter quickly attaches
  • Dual bladders and deck combine to contain 176 gallons of liquid
  • Pass-through-ports in rear of shelter allow for contaminated clothing discharge
  • All components fit into deck for easy storage and transport
  • Comes with a slip-resistant surface
  • Dimensions: 132.08 × 155.58 × 14.61cm (52 × 61.25 × 5.75 in.)
  • Weight: 55.38kg (122 lb.)
  • Weight Capacity: 2721.6kg (6000 lb.)
  • Containment Capacity: 582.95L (154 gal.)

 

Specifications

Capacity (English) 176 gal.
Material HDPE
Type Decontamination Deck
For Use With (Application) Used as an alternative to a containment pool that is used in response to any hazardous, decontamination situations
Includes Decon deck™ with two bladder attachments, 12 x 14 ft. ground tarp, gross rinse shower unit, spray wand (7gpm output), privacy shelter and supply hose
Load Bearing Capacity (English) 6000 lb.
Load Bearing Capacity (Metric) 2722 kg
Model Hospital
Dimensions (L x W x H) 61.25 x 52 x 5.75 in. (155.6 x 132.08 x 14.6 cm)
Length (English) 61.25 in.
Length (Metric) 155.6 cm
Width (English) 52 in.
Width (Metric) 132.08 cm
Height (English) 5.75 in.
Height (Metric) 14.6 cm
Weight (English) 122 lb.
Weight (Metric) 55 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.