Learn More
MSC Youngstown FR Leather, A4 Cut, 25 Cal Arc, 1 Pair
Catalog No. 5000801988
Encompass
This high-quality goatskin glove features an ergonomic, 3D form-fit pattern with a cut-resistant liner throughout the entire glove. A second layer of leather on the fingertips, palm, and knuckle reinforces durability and protection while maintaining high dexterity. These gloves will not drip, melt, or ignite when exposed to flames or sparks.
  • Cut resistant liner provides 360° protection
  • Will not drip, melt, or ignite when exposed to sparks or flames
  • Double-layered leather reinforces fingers, thumb, saddle, and palm
  • Ergonomic, 3D performance pattern form-fits to hand for superior comfort and dexterity
Catalog No. 50-008-01988 Supplier MSC Supplier No. 38032637
This high-quality goatskin glove features an ergonomic, 3D form-fit pattern with a cut-resistant liner throughout the entire glove. A second layer of leather on the fingertips, palm, and knuckle reinforces durability and protection while maintaining high dexterity. These gloves will not drip, melt, or ignite when exposed to flames or sparks.
  • Cut resistant liner provides 360° protection
  • Will not drip, melt, or ignite when exposed to sparks or flames
  • Double-layered leather reinforces fingers, thumb, saddle, and palm
  • Ergonomic, 3D performance pattern form-fits to hand for superior comfort and dexterity