The FR Mechanics Hybrid is a flame-resistant work glove offering superior dexterity, comfort, durability, and protection. Featuring a goatskin palm with 100% Kevlar® fiber by DuPont™ lining for FR, cut, and puncture resistance, and an inherently FR Nomex® fiber by DuPont™ top for flexibility. Its ergonomic 3D design provides comprehensive hand protection.
- Arc rating: 15 cal/cm²
- Cut resistance: ANSI/ISEA 105 Level A4 (palm) and A2 (top)
- Puncture resistance: ANSI/ISEA 105 Level 4
- Flame resistant: Exceeds ASTM F1358; will not drip, melt, or ignite
- Compliant: NFPA 70e-2021 / OSHA 1926
- Palm: Goat grain leather with aramid fiber lining
- Top: Inherently FR meta-aramid fiber
- Ergonomic 3D design
- Highly durable
- Applications: Gas/electric utilities, linemen, oil/gas, welding, construction, electricians