Learn More
MSC Youngstown Glove, FR Leather, Cut A4, Arc 25, 1 Pair
Catalog No. 5000803073
Encompass
This high-quality goatskin utility glove features an ergonomic, 3D form-fit pattern. It includes a cut-resistant liner for 360° protection and double-layered leather on fingertips, palm, and knuckle for enhanced durability. The glove resists dripping, melting, or ignition when exposed to flame.
  • Cut resistant liner for 360° protection
  • Flame resistant; will not drip, melt, or ignite
  • Double-layered leather on fingertips, palm, knuckle
  • Ergonomic, 3D pattern
  • Exceeds flame resistant standards (ASTM F1358)
  • ANSI/ISEA 105 Level A4 cut resistance
  • ANSI/ISEA 105 Level 5 puncture resistance
  • Arc rated 25 cal/cm² (ASTM 2675)
  • ANSI Z49.1 compliant for welding
  • NFPA 70e-2018 and OSHA 29 CFR Part 1926 compliant
Catalog No. 50-008-03073 Supplier MSC Supplier No. 05478201
This high-quality goatskin utility glove features an ergonomic, 3D form-fit pattern. It includes a cut-resistant liner for 360° protection and double-layered leather on fingertips, palm, and knuckle for enhanced durability. The glove resists dripping, melting, or ignition when exposed to flame.
  • Cut resistant liner for 360° protection
  • Flame resistant; will not drip, melt, or ignite
  • Double-layered leather on fingertips, palm, knuckle
  • Ergonomic, 3D pattern
  • Exceeds flame resistant standards (ASTM F1358)
  • ANSI/ISEA 105 Level A4 cut resistance
  • ANSI/ISEA 105 Level 5 puncture resistance
  • Arc rated 25 cal/cm² (ASTM 2675)
  • ANSI Z49.1 compliant for welding
  • NFPA 70e-2018 and OSHA 29 CFR Part 1926 compliant