Learn More
MSC Youngstown Glove, Leather Utility Plus, 20.8 cal/cm2, 2X-Large, 1 ea
Catalog No. 5000802245
Encompass
The Leather Utility Plus arc flash glove is designed for electric utility workers, offering protection and durability. Made from specially tanned goatskin grain leather, it provides a strong grip, abrasion resistance, and excellent comfort with an ergonomic, form-fitting design. The glove will not drip, melt, or ignite when exposed to sparks or flames.
  • Arc rating of 20.8 cal/cm² according to ASTM 2675
  • ANSI/ISEA 105 Level 4 puncture resistance
  • Compliant with NFPA 70e-2018 Standard and OSHA 29 CFR Part 1926
  • Constructed from specially tanned goatskin grain leather
  • Ergonomic, 3D performance pattern for a form-fit
  • Double layer leather on fingertips, knuckle, saddle, and palm for reinforcement
  • Features a 4-inch safety cuff for extended wrist protection
Catalog No. 50-008-02245 Supplier MSC Supplier No. 51639904
The Leather Utility Plus arc flash glove is designed for electric utility workers, offering protection and durability. Made from specially tanned goatskin grain leather, it provides a strong grip, abrasion resistance, and excellent comfort with an ergonomic, form-fitting design. The glove will not drip, melt, or ignite when exposed to sparks or flames.
  • Arc rating of 20.8 cal/cm² according to ASTM 2675
  • ANSI/ISEA 105 Level 4 puncture resistance
  • Compliant with NFPA 70e-2018 Standard and OSHA 29 CFR Part 1926
  • Constructed from specially tanned goatskin grain leather
  • Ergonomic, 3D performance pattern for a form-fit
  • Double layer leather on fingertips, knuckle, saddle, and palm for reinforcement
  • Features a 4-inch safety cuff for extended wrist protection