Learn More
MSC Youngstown Kevlar Lined Gloves, ANSI Cut A4, Large, 1 Pair
Catalog No. 5000685273
Encompass
These utility gloves are made with Kevlar as the primary and lining material, offering cut, puncture, and abrasion resistance. Featuring a black and gray design, they meet ANSI/ISEA standards for Cut Resistance Level A4, Puncture Resistance Level 4, and Abrasion Resistance Level 4, and are designed for industrial applications.
  • Supportive cuff with adjustable Velcro closure
  • Three-dimensional glove design pattern
  • Terry cloth brow wipe
  • Interior padded protection
  • Machine washable and air dry
Catalog No. 50-006-85273 Supplier MSC Supplier No. 75994301
These utility gloves are made with Kevlar as the primary and lining material, offering cut, puncture, and abrasion resistance. Featuring a black and gray design, they meet ANSI/ISEA standards for Cut Resistance Level A4, Puncture Resistance Level 4, and Abrasion Resistance Level 4, and are designed for industrial applications.
  • Supportive cuff with adjustable Velcro closure
  • Three-dimensional glove design pattern
  • Terry cloth brow wipe
  • Interior padded protection
  • Machine washable and air dry