Learn More
Teknova YPAD Agar Plates with 0.01% Adenine Hemisulfate. 100mm plates, 20 plates per sleeve, Sterile. 4 pack.
SDP
Catalog No. 502387253
Encompass
1.00% Yeast Extract, 2.00% Peptone,2.00% Agar, 2.00% Cerelose Anhydrous Dextrose (Glucose), 0.01% Adenine Hemisulfate
Catalog No. 50-238-7253 Supplier Teknova Supplier No. Y22254PK
1.00% Yeast Extract, 2.00% Peptone,2.00% Agar, 2.00% Cerelose Anhydrous Dextrose (Glucose), 0.01% Adenine Hemisulfate