Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp YPD Agar, 1 Gram Capsules, 500 G
SDP

Catalog No. 501977684
Encompass_Preferred

Mix 65 capsules in one liter distilled water until evenly dispersed. Heat with repeated stirring and boil for one minute to dissolve completely. Distribute and autoclave at 121°C for 15 minutes.

Ingredients:

Peptone: 20 g/l
Dextrose: 20 g/l
Yeast Extract: 10 g/l
Agar: 15 g/l

Catalog No. 50-197-7684 Supplier Research Products International Corp Supplier No. Y20085500.0
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mix 65 capsules in one liter distilled water until evenly dispersed. Heat with repeated stirring and boil for one minute to dissolve completely. Distribute and autoclave at 121°C for 15 minutes.

Ingredients:

Peptone: 20 g/l
Dextrose: 20 g/l
Yeast Extract: 10 g/l
Agar: 15 g/l

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.