Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp YPD Agar, 1 Kilogram
SDP

Catalog No. 50489165
Encompass_Preferred

For maintaining and propagating yeasts in molecular biology applications. The addition of protein and yeast extract promotes better growth.

Add 65 grams per liter of water. Heat and stir until completely dissolved. Sterilize by autoclaving for 15 minutes.

Ingredients:

Peptone: 20 g/L
Dextrose: 20 g/L
Yeast Extract: 10 g/L
Agar: 15 g/L

Catalog No. 50-489-165 Supplier Research Products International Corp Supplier No. Y200801000.0
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For maintaining and propagating yeasts in molecular biology applications. The addition of protein and yeast extract promotes better growth.

Add 65 grams per liter of water. Heat and stir until completely dissolved. Sterilize by autoclaving for 15 minutes.

Ingredients:

Peptone: 20 g/L
Dextrose: 20 g/L
Yeast Extract: 10 g/L
Agar: 15 g/L

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.