Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp YPD Agar, 333ml Capsules, 500 Grams
SDP

Catalog No. 50489170
Encompass_Preferred

Mix 3 capsules in 1 liter of purified water until evenly dispersed. Heat with repeated stirring and boil for 1 minute to dissolve completely. Sterilize by autoclaving at 121°C for 15 minutes.

Ingredients:

Peptone: 20 g/l
Dextrose: 20 g/l
Yeast Extract: 10 g/l
Agar: 15 g/l

Catalog No. 50-489-170 Supplier Research Products International Corp Supplier No. Y20086500.0
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mix 3 capsules in 1 liter of purified water until evenly dispersed. Heat with repeated stirring and boil for 1 minute to dissolve completely. Sterilize by autoclaving at 121°C for 15 minutes.

Ingredients:

Peptone: 20 g/l
Dextrose: 20 g/l
Yeast Extract: 10 g/l
Agar: 15 g/l

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.