Learn More
Teknova YPD Agar Plates. 150mm Plates, 20 Plates per Sleeve, Sterile. 4 pack.
SDP
Supplier:  Teknova Y10014PK
Encompass
YP (Yeast Peptone) media is a growth medium used to culture yeast. YP is a rich source of proteins and nutrients essential for the development of yeast cells.
Catalog No. 50-238-7688