Learn More
Teknova YPD Agar Plates with 1M Sorbitol and ClonNat-100. 100mm, 20 Plates Sleeve, Sterile. 8 pack.
SDP
Catalog No. 502745934
Encompass
YP (Yeast Peptone) media is a growth medium used to culture yeast. YP is a rich source of proteins and nutrients essential for the development of yeast cells.
Catalog No. 50-274-5934 Supplier Teknova Supplier No. Y16638PK
YP (Yeast Peptone) media is a growth medium used to culture yeast. YP is a rich source of proteins and nutrients essential for the development of yeast cells.