Learn More
Teknova YPD Agar Plates with 1M Sorbitol and Geneticin G418-1000. 100mm, 20 plates/sleeve. Sterile. 8 pack.
SDP
Catalog No. 502745936
Encompass
YP (Yeast Peptone) media is a growth medium used to culture yeast. YP is a rich source of proteins and nutrients essential for the development of yeast cells.
Catalog No. 50-274-5936 Supplier Teknova Supplier No. Y16618PK
YP (Yeast Peptone) media is a growth medium used to culture yeast. YP is a rich source of proteins and nutrients essential for the development of yeast cells.