Learn More
Teknova YPD Agar Plates with Animal-Free Soytone. 100mm, 20 Plates per Sleeve, Sterile. 4 pack.
SDP
Catalog No. 502387727
Encompass
YP (Yeast Peptone) media is a growth medium used to culture yeast. YP is a rich source of proteins and nutrients essential for the development of yeast cells.
Catalog No. 50-238-7727 Supplier Teknova Supplier No. Y10054PK
YP (Yeast Peptone) media is a growth medium used to culture yeast. YP is a rich source of proteins and nutrients essential for the development of yeast cells.