Learn More
Teknova YPD Broth. 15Kg.
SDP
Catalog No. 50843583
Encompass
YP (Yeast Peptone) media is a growth medium used to culture yeast. YP is a rich source of proteins and nutrients essential for the development of yeast cells.
Catalog No. 50-843-583 Supplier Teknova Supplier No. Y5301
YP (Yeast Peptone) media is a growth medium used to culture yeast. YP is a rich source of proteins and nutrients essential for the development of yeast cells.