Learn More
Research Products International Corp YPD Broth, 5 Kilograms
SDP
Catalog No. 50489173
Encompass_Preferred
For maintaining and propagating yeasts in molecular biology applications. The addition of protein and yeast extract promotes better growth.
Dissolve 50 grams in 1 liter of distilled water. Sterilize by autoclaving 15 minutes.
Ingredients:
Peptone: 20 g/l
Dextrose: 20 g/l
Yeast extract: 10 g/l
Catalog No. 50-489-173 Supplier Research Products International Corp Supplier No. Y200905000.0
For maintaining and propagating yeasts in molecular biology applications. The addition of protein and yeast extract promotes better growth.
Dissolve 50 grams in 1 liter of distilled water. Sterilize by autoclaving 15 minutes.
Ingredients:
Peptone: 20 g/l
Dextrose: 20 g/l
Yeast extract: 10 g/l